Super Erecta® Stem Casters, Metro™
Supplier: Metropolitan Wire
4LD 5M 5MB 5MPR 5LD 5MDBGSA 5MDGSA 5MR 5MFA 5MFBA 5MDA 5MESD 5MBESD 5MDRA 5MC 5MBC 5MDRGSA 5MDBA 6MP 6MPB 6MPR 5MP
CA19766-236EA 89.02 CAD
CA19766-236 CA19766-238 CA19766-240 CA19766-250 CA19766-474 CA19766-480 CA19766-482 CA19766-492 CA19766-792 CA19766-794 CA19766-816 CA19767-004 CA19767-006 CA22221-188 CA22221-198 CA22221-200 CA22221-204 CA22221-208 CA97005-486 CA97005-488 CA97005-490 CA19766-244
Super Erecta® Stem Casters, Metro™
Furniture Shelves
For use with Super Erecta® shelving and SiteSelect® posts to create mobile units. Swivel, foot-operated brake, and rigid styles are available.
  • Resilient rubber tread casters are non-marking and feature a molded, soft tread that provides quiet operation
  • Polyurethane tread casters are abrasion-resistant, non-marking, and shock-absorbant
Casters with high modulus rubber tread provide extra shock protection and have excellent resistance to oil and grease. Special cart-washable casters are constructed of stainless steel components. Casters include greaseMore Product Information