HD Super™ Mobile Worktables, Metro™
Supplier: Metropolitan Wire
MWT305FS MWT305US MWT306US MWT307FS MWT309FS MWT306HS MWT307HS MWT309HS MWT306FS MWT307US MWT309US
CA82000-960EA 4564.66 CAD
CA82000-960 CA82000-962 CA82020-026 CA97005-372 CA97005-764 CA97005-766 CA97005-768 CA97005-770 CA97005-772 CA97005-924 CA97005-926 CA97005-928 CA97005-930 CA97005-932
HD Super™ Mobile Worktables, Metro™
Furniture Tables and Workbenches
Exceptionally stable, easy-to-clean conductive tables are ideal for the cleanroom, classroom, or laboratory.
  • Seamless 14-gauge Type 304 stainless steel work surface with a support structure that virtually eliminates drumming
  • Fitted with four 12.7 cm (5") diameter polyurethane swivel casters, two with wheel brakes
  • Work surface fits over plastic sleeves at the top of the posts
Tables are available with either a bottom shelf, a three-sided horizontal tubular frame support, or a horizontal "H" frame support. Shelves are adjustable in 5.1 cm (2")More Product Information