InLab® Power Combination pH Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
51343110
CA97001-620EA 1621.65 CAD
CA97001-620
InLab® Power Combination pH Electrode, METTLER TOLEDO®
Electrodes pH Electrodes
Electrode features integrated SteadyForce™ reference system with under-over pressure to guarantee electrode discharge and prevent contamination
  • Integrated SteadyForce™ reference system
  • Ceramic junction
  • Constant, controlled discharge
  • A41 body
  • Resistant to corrosives
Ceramic junction allows for constant and controlled discharge of electrolyte. Special A41 glass body is resistant to corrosive or organic samples and is ideal for high temperatures and biological media. S7 screw head connects with different cables, so electrode may be used with a wide range of meters.
Order Now
SPECIFICATIONS
Environmentally Preferable
Ø