MAbTrap Column Kit, Cytiva
Supplier: Cytiva
17-1128-01
CA95056-072LEA 1388.83 CAD
CA95056-072L
MAbTrap Column Kit, Cytiva
Chromatography Columns
MAbTrap Kit is an affinity chromatography kit for fast and effective purification of monoclonal and polyclonal IgG from ascites fluid, serum, and cell culture supernatants.
  • Convenient kit for purification of IgG from monoclonal and polyclonal material using a syringe in less than 15 min
  • MAbTrap Kit contains everything necessary to perform a complete one-step purification without using complex equipment.
MAbTrap Kit contains everything necessary to perform a complete one step purification without using complicated equipment. It also eliminates time-More Product Information