GSTrap 4B Columns

Supplier: Cytiva
28-4017-47 28-4017-48 28-4017-45 29-0486-09
CA95056-050LEA 960.76 CAD
CA95056-050L CA95055-968L CA95056-278L CA11033-014
GSTrap 4B Columns
Chromatography Columns
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins.

  • High binding capacity
  • Mild elution conditions preserving protein antigenicity and function
  • Easy one-step purifications of Glutathione S-Transferase (GST) tagged proteins resulting in high purity
  • GSTrap 4B are designed to be used with a syringe, pump, or chromatography system, such as ÄKTA design

GST-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrapMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. pressure 5 bar [0.5 MPa] (70 psi)
pH range 4 - 13
Storage 4 to 30 °C, 20% Ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR