Sepharose™ 6B Gel Filtration Base Matrix
Supplier: Cytiva
17-0110-01
CA95056-026LEA 2499.03 CAD
CA95056-026L
Sepharose™ 6B Gel Filtration Base Matrix
Protein Purification Protein Purification Resins
Sepharose™ 6B is an agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
  • 6% agarose gel filtration media
  • Proven base matrix for coupling affinity ligands