HiTrap Columns, rProtein A FF, Cytiva
Supplier: Cytiva
17-5079-02 17-5080-01 17-5079-01 17-5080-02
CA95055-868LPK 1029.88 CAD
CA95055-868L CA95056-008L CA95056-088L CA95056-090L
HiTrap Columns, rProtein A FF, Cytiva
Chromatography Columns
HiTrap rProtein A FF columns are prepacked with rProtein A Sepharose Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources, including cell supernatants and ascites.
  • Quick, convenient preparative purification of monoclonal antibodies
  • Well-established protein A-based resin used in many approved MAb processes
  • Simple operations with a syringe, pump, or high-performance chromatography system, such as ÄKTA design
Recombinant protein A exhibits similar Fc region specificity to that of native protein A but showsMore Product Information