PE SOURCE 15Q Columns, Cytiva
Supplier: Cytiva
17-5181-01
CA95055-900LEA 3275.3 CAD
CA95055-900L
PE SOURCE 15Q Columns, Cytiva
Chromatography Columns
SOURCE™ 15Q PE is a strong anion exchanger prepacked in a Tricorn column for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
  • Improved stability in organic solvents and pH extremes
  • Fast, high-resolution purifications and easy scale-up
  • Improved capacity compared to MonoBeads but with a slightly lower resolution
Rigid, monodispersed, spherical particles with controlled pore-size of 15 µm offers excellent flow characteristics and sample loading of up to 25 mg of protein/ml.