HiTrap Columns, HiTrap HIC Selection Kit, Cytiva
Supplier: Cytiva
28-4110-07
CA95055-860LPK 1013 CAD
CA95055-860L
HiTrap Columns, HiTrap HIC Selection Kit, Cytiva
Chromatography Columns
HiTrap HIC Selection Kit is designed for rapid screening for the most appropriate hydrophobic interaction chromatography (HIC) resin and experimental conditions.
  • High likelihood of finding suitable HIC resin – contains seven different resins
  • Convenient prepacked HiTrap columns, compatible with syringe, single laboratory pump, or chromatography systems such as ÄKTA systems
Within HIC you need to perform empirical testing to find optimal conditions for your target sample. The parameters you typically screen for are type of salt, concentration of salt,More Product Information