Glass Plates, Cytiva
Supplier: Cytiva
80-6178-99
CA95044-738PK 247.92 CAD
CA95044-738
Glass Plates, Cytiva
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The key to reproducible electrophoresis results is temperature control. Gels cooled on their two faces produce slanted bands appearing as broad or double bands in stained gels. SE 600 Ruby ensures uniform heat in gels because, the gels are immersed in the lower buffer for uniform heat transfer and a standard magnetic stirrer circulates buffer around the vertically suspended heat exchanger to maintain homogeneous lower buffer temperature. The ergonomic SE 600 Ruby provides two built-in side handles, which facilitates carrying and handling. Run up to four gels (up to 112 samples) in a singleMore Product Information