Extraction Reagents 2 (5X), (10 ml), Enzo Life Sciences
Supplier: Enzo Life Sciences
80-1502
CA95043-696EA 244.9 CAD
CA95043-696
Extraction Reagents 2 (5X), (10 ml), Enzo Life Sciences
Buffers Extraction Buffers
Concentrated lysis buffer for preparation of cell and tissue extracts.
TRIS buffer containing detergents.