Ammonium persulfate (APS) 150 mg, tablets for electrophoresis

Supplier: G-Biosciences
Danger

786-510
CA95043-468EA 930.08 CAD
CA95043-468
Ammonium persulfate (APS) 150 mg, tablets for electrophoresis
Ammonium peroxodisulphate (APS)

G-Biosciences offers Ammonium Persulfate (APS), the catalyst for the polymerization of polyacrylamide gels, in the form of ready-to-use tablets (100 count) or as a powder (100 g)


For the tablets, simply add 1 tablet to 1.5 mL ultra pure water for a 10% solution.

Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR