UVP Transilluminator PLUS, Analytik Jena
Supplier: Analytik Jena CA
95-0422-01 95-0422-02 95-0424-01 95-0424-02 95-0421-01 95-0421-02 95-0427-01 95-0427-02 95-0428-02 95-0429-02 95-0430-01 95-0430-02 95-0431-01 95-0431-02 95-0432-01 95-0432-02 95-0425-01 95-0425-02 95-0426-01 95-0420-01 95-0420-02 95-0423-01 95-0423-02
CA95041-534EA 8452.15 CAD
CA95041-534 CA95041-536 CA95041-538 CA95041-540 CA95041-542 CA95041-544 CA95041-546 CA95041-548 CA95041-552 CA95041-556 CA95041-558 CA95041-560 CA95041-562 CA95041-564 CA95041-566 CA95041-568 CA95041-570 CA95041-572 CA95041-574 CA95042-398 CA95042-400 CA95042-402 CA95042-404
UVP Transilluminator PLUS, Analytik Jena
Transilluminators UV Transilluminators
High performance 25 watt UV tubes for high fluorescence and sensitivity on stained gels
Caution: Transilluminators are powerful sources of ultraviolet radiation which can cause serious damage to unprotected eyes and skin. Read the ultraviolet Caution Information before operating any transilluminators.Caution: Transilluminators are powerful sources of ultraviolet radiation which can cause serious damage to unprotected eyes and skin. Read the ultraviolet Caution Information before operating any transilluminators.
The stainless steel cover frame provides an easy clean surface and reducesMore Product Information