UVP UV/White Light Transilluminators, Analytik Jena
Supplier: Analytik Jena CA
95-0415-01 95-0415-02 95-0415-04 95-0415-05 95-0418-01 95-0418-02
CA95041-578EA 6058.12 CAD
CA95041-578 CA95041-580 CA95041-582 CA95041-584 CA95041-586 CA95041-588
UVP UV/White Light Transilluminators, Analytik Jena
Transilluminators UV/White Light Transilluminators
UV/White Transilluminators house white light and UV working areas side by side for multiple applications
  • Increased ultraviolet output and intensity
  • No light flicker
  • Wider intensity range with variable selection (select models)
  • Fast lamp start up
  • Universal power supply
  • Reduced electrical consumption
  • Longer tube life
  • Lighter weight
  • Improved gel visualization and gel documentation
Caution: Transilluminators are powerful sources of ultraviolet radiation which can cause serious damage to unprotected eyes and skin. Read the ultraviolet Caution InformationMore Product Information