Potassium-D-gluconate, anhydrous ≥97% USP

Supplier: Spectrum Chemical Mfg. Corp.

PO178-500GM
CA95035-160EA 351 CAD
CA95035-160
Potassium-D-gluconate, anhydrous ≥97% USP
Potassium D-gluconate

Potassium Gluconate, Anhydrous, USP is a mineral supplement. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄COOK
Melting Pt: 174…176 °C
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064211
CAS Number: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR