Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Supplier: Simport Scientific
M517-2SL M517-3SL M517-4SL M517-5SL M517-6SL M517-7SL M517-8SL M517-9SL M517-10SL M517-11SL M517-12SL M517-16SL M517-17SL M517-18SL M518-2SL M518-3SL M518-4SL M518-5SL M518-6SL M518-7SL M518-8SL M518-9SL M518-10SL M518-11SL M518-12SL M518-16SL M518-17SL M518-18SL
CA95030-286CS 289.36 CAD
CA95030-286 CA95030-288 CA95030-290 CA95030-292 CA95030-294 CA95030-296 CA95030-298 CA95030-300 CA95030-302 CA95030-304 CA95030-306 CA95030-308 CA95030-310 CA95030-312 CA95030-314 CA95030-316 CA95030-318 CA95030-320 CA95030-322 CA95030-324 CA95030-326 CA95030-328 CA95030-330 CA95030-332 CA95030-334 CA95030-336 CA95030-338 CA95030-340
Slimsette™ Processing Cassettes in QuickLoad™ Sleeves, Simport Scientific
Embedding Cassettes Cassettes for Labeling Instruments
These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle. All cassettes colors are non-cytotoxic and non-metallic. Dimensions: 42.2Lx28.5Wx6Hmm (121/32x11/More Product Information