Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
786-025 786-029 786-029G R025 786-420 786-421 786-472 786-474 786-475 786-479 786-480 786-481 786-482 786-483 786-484 786-485 786-486 786-060 786-531 786-532
CA82021-242EA 100.66 CAD
CA82021-242 CA82021-250 CA82021-252 CA82021-496 CA95029-224 CA95029-226 CA95029-250 CA95029-252 CA95029-254 CA95029-262 CA95029-264 CA95029-266 CA95029-268 CA95029-270 CA95029-272 CA95029-274 CA95029-276 CA89167-720 CA95043-524 CA95043-526
Electrophoresis Running Buffers, G-Biosciences
Buffers Electrophoresis Buffers Running Buffers
G-Biosciences offers many electrophoresis running buffers including: TAE Running Buffer [50X], TBE Running Buffer [10X], TBE-Urea Sample Buffer, MOPS Running Buffer [10X], and MOPS/ SDS Running Buffer [20X]
  • The TAE Running Buffer [50X] contains 2M Tris-Acetate and 0.05M EDTA (at a pH of 8.3)
  • The TBE Running Buffer [10X] contains 0.9M Tris-borate and 0.01M EDTA (at a pH of 8.3)
  • The MOPS Running Buffer [20X] contains 200 mM MOPS (at a pH of 7.0), 80 mM Sodium acetate and 10 mM EDTA
G-Biosciences' TBE-Urea Sample Buffer is a loading buffer used to resolve single-stranded DNA oligos or RNA into sharp, distinct bands. The TBE-Urea Sample Buffer is for the analysis and purification of products ranging from 20-800 bases, making them an ideal choice for synthetic oligo analysis and purification, RNase Protection Assays (RPA), in vitro transcription studies, and Northern blot analysis.
These include the SDS-PAGE Sample Loading Buffer, SDS-PAGE Running Buffer, Native Sample Loading Buffer, Tris Glycine Native Gel Running Buffer, Tricine Sample Buffer, Tris-Tricine, Tris-Tricine-SDS, MES SDS Running Buffer, MOPS/ SDS Running Buffer, IEF Anode Buffer, IEF Cathode Buffer, Zymogram Sample Buffer, Zymogram Renature Buffer, Zymogram Development Buffer and the Discontinuous Buffer System.