illustra™ GFX™ PCR DNA and Gel Band Purification Kits

Supplier: Cytiva

28-9034-70 28-9034-71
CA95026-728LEA 668.85 CAD
CA95026-728L CA95026-730L CA101725-244 MPCA29489379
illustra™ GFX™ PCR DNA and Gel Band Purification Kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The illustra GFX PCR DNA and Gel Band Purification kits are for the isolation and concentration of DNA fragments from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction enzyme digests.


  • Fast and easy-to-use method with less than 10 min hands-on time
  • Flexible 10 to 50 μl elution volume for different DNA concentration needs
  • Improved column design with increased maximum volume
  • Holding of up to 800 μl liquid volume (950 μl without a cap)
  • Two choices of elution buffer provided to fit downstream applications
  • Color-coded caps for ease of use


illustra GFX PCR DNA and Gel band purification kit is designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg. The kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin column for the purification of DNA from both solution and agarose gel. Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution. DNA purity is exceptional, 99.5% of contaminants are removed.


Highly pure DNA is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning. Addition of a visual color indicator to the capture buffer ensures optimal pH for maximum DNA binding and recovery.


Ordering information: Kit contains GFX columns (2×50), collection tubes (2×50). color-coded bottles of capture buffer (blue, 55 ml), wash buffer (yellow, 25 ml), and two elution buffers each 12 ml (Tris-HCl and sterile water) and an instruction booklet.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule DNA
Refine target molecule DNA gel bands,Amplified/modified DNA
Sample type Gel matrix,PCR and other enzymatic reactions
Method / Format Column (silica-based)
Prep size Mini
Sample size 50 bp - 10 kb
Yield PCR 10 µl elution volume - 65%, PCR 50 µl elution volume - 82%, 300 mg agarose 10 µl elution volume - 57%, 300 mg agarose 50 µl elution volume - 91%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR