PA Coliform Kit, HiMedia®
Supplier: HIMEDIA LABORATORIES PVT. CA
MS1186-10KT
CA95022-750EA 1127.54 CAD
CA95022-750
PA Coliform Kit, HiMedia®
Media for Microbiology Dehydrated Media for Microbiology
The kit is used for the detection of presence and absence of coliform bacteria in water from treatment plants or distribution systems.
Delivery information: The kit contains dehydrated medium (3x concentration), sterile bottle (1 mL capacity) and ziplag bag (1).
Caution: Store below 30 °C. The kit has shelf life of 2 years.