Enriched Thioglycollate Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M738-500G
CA95022-200EA 1315.54 CAD
CA95022-200
Enriched Thioglycollate Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation, cultivation and identification of a wide variety of obligate anaerobic bacteria.
  • Microbiological culture media
  • 500 g poly bottles