Caffeic Acid Ferric Citrate Test Agar Dehydrated Culture Media, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M563-500G
CA95022-002EA 3458.57 CAD
CA95022-002
Caffeic Acid Ferric Citrate Test Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation, presumptive identification, and differentiation of Cryptococcus neoformans from other species.
  • Microbiological culture media
  • 500 g poly bottles