ISP Medium No. 2 Dehydrated Culture Media, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M424-500G
CA95021-804EA 1153.81 CAD
CA95021-804
ISP Medium No. 2 Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and cultivation of yeasts, molds, and other aciduric microorganisms.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
SPECIFICATIONS
Environmentally Preferable