Water Molecular_Biology, sterile

Supplier: HIMEDIA LABORATORIES PVT. CA

Synonyms: Hydrogen oxide

TCL016-500ML
CA95016-376PK 1919.24 CAD
CA95016-376 CA95016-374
Water Molecular_Biology, sterile
Water

This water is molecular biology grade.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR