CP-710 Non-Hazardous Alkaline Detergent, CANI
Supplier: Cani
CAN-710-15 CAN-710-275 CAN-710-30 CAN-710-55 CAN-710-5 CAN-710-1 CAN-710-2.5
CA93001-306EA 3398.99 CAD
CA93001-306 CA93001-310 CA93001-312 CA93001-314 CA93001-316 CA93001-304 CA93001-308
CP-710 Non-Hazardous Alkaline Detergent, CANI
Cleaning Agents Detergents and Surface Cleansers
Concentrated liquid formulated for the pharmaceutical, biotechnology, cosmetic, and medical device industries
Effectively removes a wide range of process and research residues, including serum, proteins, organics, and oils. Safe for use on aluminum, galvanized and soft metals, glass, stainless steel, and a wide variety of plastics. Ideal for laboratory animal cage wash applications at 0.2–1% use dilution. Designed to be low-foaming at all temperatures for easy rinsing and will not cavitate recirculation pumps. Free-rinsing formulation provides clean, residue-free surfaces. ChelantsMore Product Information