BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
260673 260678 260683 260684 260001 260671 260672 260674 260679 260680 260685 260003 260002
CA90003-638EA 243.64 CAD
CA90003-638 CA90003-642 CA90003-648 CA90003-650 CA90010-202 CA90003-634 CA90003-636 CA90003-640 CA90003-644 CA90003-646 CA90003-652 CA75805-552 CA75805-942
BD GasPak™ EZ Container Systems, BD Diagnostics
Anaerobic Systems
Rapid gas generating sachets produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments.
Pouch systems include 20 resealable pouches and 20 rapid gas-generating sachets. The Anaerobe Pouch System also includes 20 dry anaerobic indicators. Sachets do not require water or a catalyst to produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments. Pouches are one-step resealable; no sealing bars needed. Multi-layer plastic pouch material preserves environmental integrity for maximumMore Product Information