Arium® Comfort II Type 1 and 2 Combined Water Systems
Supplier: Sartorius
H2O-II-2-B H2O-II-2-T H2O-II-2-UV-T H2O-II-2-UV-B H2O-II-2-TOC-B H2O-II-2-TOC-T H2O-II-2-B H2O-II-2-T H2O-II-2-UV-T H2O-II-2-UV-B H2O-II-2-TOC-B H2O-II-2-TOC-T H2O-II-1-T H2O-II-1-UV-T H2O-II-1-TOC-T H2O-II-1-UV-B H2O-II-1-TOC-B H2O-II-1-B
89510-600EA 31291.09 CAD
89510-600 89510-604 89510-606 89510-610 89510-612 89510-616 CA89510-600 CA89510-604 CA89510-606 CA89510-610 CA89510-612 CA89510-616 75995-098 75995-100 75995-102 75995-104 75995-106 75995-108
Arium® Comfort II Type 1 and 2 Combined Water Systems
Water Purification Systems Pure and Ultra Pure Water Systems
The arium comfort systems produce ASTM Type 1 ultrapure water and Type 2 pure water.
  • Efficient iJust Technology
  • Unique Filter Cartridge
  • Touch-Screen Control Panel
  • UV Lamp Prevents Microbiological Growth
  • Systems Produce ASTM Type 1 and 2 Water
The systems feature the latest reverse osmosis technology combined with electrodeionization (EDI) and a unique filter cartridge for producing ultrapure water of the highest quality, delivering rates up to 120 L per hour
The systems also feature efficient iJust technology, flexible and space-saving twinMore Product Information