Celite® 503, filter aid, Supelco®

Supplier: MilliporeSigma
Danger

1.02692.1000 1.02692.1000
89501-600EA 497.05 CAD
89501-600 CA89501-600
Celite® 503, filter aid, Supelco®
Celite® 503, filter aid
Formula: O₂Si
CAS Number: 68855-54-9

Order Now

Specification Test Results

Solubility  - 0.001 g/l (20 °C)
Bulk Density 300 kg/m3
pH 10 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR