Surface Water Test Kit, Hach
Supplier: Hach
2559800
89500-750EA 1175.22 CAD
89500-750 CA89500-750
Surface Water Test Kit, Hach
Environmental Test Kits
The ideal tool for source water monitoring and wastewater treatment plants, this kit uses drop count titration and color disc methods to provide economical, easy-to-use tests for ammonia, chlorine, pH, nitrate, dissolved oxygen, phosphorus, and temperature
  • Ideal tool for both industrial and municipal applications
  • Uses drop count titration and color disc methods to provide economical, easy-to-use tests for ammonia, chlorine, pH, titrate, dissolved oxygen, phosphorus, and temperature
  • Rugged, lightweight kit includes the convenient pH Pocket Pal™ Tester and all necessary apparatus and reagents
Caution: Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
The rugged, lightweight kit includes the convenient pH Pocket Pal™ Tester and all necessary apparatus and reagents for testing.
Ordering information: Includes ten reagents, deionized water, color discs, thermometer, tubes, pH tester, clippers, dropper, measuring tube, bottle, BOD bottle, color comparator box, viewing adapter, instruction sheet, and carrying case.