SpectraMax® M2/M2e Multimode Microplate Readers, with SoftMax® Pro Software, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
M2 M2E M2 M2E M2-SC
89429-532EA 123292.82 CAD
89429-532 89429-534 10067-744 10067-746 CA89429-532 CA89429-534 CA10067-744
SpectraMax® M2/M2e Multimode Microplate Readers, with SoftMax® Pro Software, Molecular Devices
Microplate Readers
The SpectraMax® M2 and M2e Multimode Microplate Readers provide two detection modes in one platform and offer the added functionality of bottom-read fluorescence for cell-based assays.
  • Two detection modes
  • PathCheck® PathLength measurement technology
  • Eliminate standard curves
SpectraMax® M2 and M2e Microplate Readers are the standard for UV/visible multimode reader absorbance, providing ultrafast and full spectral range detection for cuvettes and 6-, 12-, 24-, 48-, 96-, and 384-well microplates
For fluorescence intensity assays, the SpectraMax® M2More Product Information