Middlebrook 7H11 Selective, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
W40
CA89429-350PK 92.95 CAD
CA89429-350
Middlebrook 7H11 Selective, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Middlebrook 7H11 Selective, for mycobacteria, deep fill, 15x100mm plate, order by the package of 10, by Hardy Diagnostics
(Mitchison formula with CAPT antibiotics) For selective isolation of Mycobacterium spp.