Bile HardyDisks™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Z7091
CA89426-240EA 189.43 CAD
CA89426-240
Bile HardyDisks™, Hardy Diagnostics
Microbiology Test Systems Endotoxin Tests
Bile HardyDisks™, to aid in the identification of Bacteroides fragilis group on the basis of bile resistance, 1 x 50 disks by Hardy Diagnostics, freeze on arrival.
Differentiates Bacteroides fragilis group from other anaerobic bacteria. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispense.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Differentiates Bacteroides fragilis group from other anaerobic bacteria
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispense.