Peptone Salt, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
U390
CA89422-670PK 560.85 CAD
CA89422-670
Peptone Salt, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
For use as a diluent and for the preparation of suspensions.
  • Diluent
  • 0.1% proteose peptone
  • 0.85% NaCl
Contains 0.1% proteose peptone and 0.85% NaCl.
Ordering information: Minimum order of 5 packs.