FC Recirculating Chillers, JULABO
Supplier: JML SCIENCES INC CA
9600060.13 9601060.13 9600063.13 9601063.13 9600160.13 9600163.13 9600166.13 9601256.13 9600060 9601060 9600063 9601063 9600160 9600163 9600166 9601256
89421-908EA 33907.13 CAD
89421-908 89421-910 89421-912 89421-914 89421-916 89421-918 89421-920 89421-922 CA89421-908 CA89421-910 CA89421-912 CA89421-914 CA89421-916 CA89421-918 CA89421-920 CA89421-922
FC Recirculating Chillers, JULABO
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Recirculating chillers with integrated heater.
  • German engineering and quality
  • Expanded temp range to 80 °C
  • Pt100 connection for FC1600T, FCW2500T
JULABO FC Series of environmentally friendly recirculating coolers save tap water and provide powerful cooling for short cool-down times.
FC units feature two separate Multi-Displays (LED) for actual and set point values. Models designated FCW are water-cooled which reduces the noise level and reduced heat generation to the laboratory environment. The units feature permanent performance supervision viaMore Product Information