Barnstead™ GenPure™ xCAD Plus Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
50136151 50136152 50136151 50136153 50136146 50136169 50136172 50136170 50136171
89414-720EA 23858 CAD
89414-720 CA89414-716 CA89414-720 CA89414-722 CA89414-724 CA89414-732 CA89414-736 CA89414-728 CA89414-734
Barnstead™ GenPure™ xCAD Plus Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead GenPure xCAD Plus water purification systems provides up to 200L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water
The GenPure xCAD Plus systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. The xCAD remote dispenser can be mounted on the wall or on the bench for maximum flexibility. In addition, up to two additional remote dispensers can be added to create up to three work stations from one water system.
The GenPure xCAD systems are offered in a variety of configurations to meet aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
W×D×H 372×330×615 mm (14.6×13×24")