AlphaTec® 58-335, Nitrile Gloves, with AQUADRI® Technology, Ansell
Supplier: Ansell Canada
114019 114020 114021 114022 114023
CA89413-064CS 1502.39 CAD
CA89413-064 CA89413-066 CA89413-068 CA89413-070 CA89413-072
AlphaTec® 58-335, Nitrile Gloves, with AQUADRI® Technology, Ansell
Gloves
Ansell’s new, proprietary AQUADRI® technology delivers the properties of a sponge with great moisture absorption capacity, featuring AquaDri® Nitrile Foam Lining, thus keeping hands drier for a longer period of time resulting in improved comfort and performance.
  • Superior puncture and abrasion protection
  • Improved dry and wet grip
  • Gauntlet cuffs prevent dripping onto forearms
  • Great moisture absorption capacity
These gloves are designed to deliver optimal results in wet or dry work environments where chemical protection is crucial.
Ideal uses areMore Product Information