Nutra-Gel Diet™, Grain-Based Formula, Rodent, Dry Mix Kit, Bio-Serv
Supplier: Bio-Serv
F4798-KIT
CA89408-234EA 224.12 CAD
CA89408-234
Nutra-Gel Diet™, Grain-Based Formula, Rodent, Dry Mix Kit, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete, grain-based, cherry-flavored gel diet that provides both food and water for rodents, and is especially useful in cases involving chewing and mobility impairments, post-op care, and weanlings
  • Kit provides the powdered diet and trays to make 4 trays of 16 cubes/tray at 40 g/cube of Nutra-Gel
  • Easy to prepare; just add hot water and mix according to directions
  • Prepare desired quantities of Nutra-Gel as needed
  • Place Nutra-Gel directly on the bedding or in the food hopper
  • Nutra-Gel remains moist in the cage 24 to 48 hours, depending on
More Product Information