Tetrathionate Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K65 K164
CA89407-968PK 338.21 CAD
CA89407-968 CA89408-136
Tetrathionate Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Salmonella species
Use with iodine-iodide solution 89407-624.