OF Medium, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Y57 Y58 Y53
CA89407-786PK 311.5 CAD
CA89407-786 CA89407-788 CA89407-790
OF Medium, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the detection of carbohydrate oxidation or fermentation by bacteria.