Kligler Iron Agar (KIA), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
L70 R70
CA89407-626PK 573.24 CAD
CA89407-626 CA89407-628
Kligler Iron Agar (KIA), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the identification of gram negative enteric bacteria based on hydrogen sulfide production and the fermentation of dextrose and lactose.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2-8°C