Fraser Broth, Modified, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
K98
CA89407-598PK 484.5 CAD
CA89407-598
Fraser Broth, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Listeria species (without ferric ammonium citrate).