Fluid Thioglycollate, USP
Supplier: HARDY DIAGNOSTICS CA
U207 U121 U434
CA89407-581PK 734.51 CAD
CA89407-581 CA89423-724 CA10124-494
Fluid Thioglycollate, USP
Microbiology Test Systems Endotoxin Tests
Fluid Thioglycollate Medium (FTM), is recommended for the use of of detecting microorganisms in sterile materials and USP sterility procedures.
Order Now
SPECIFICATIONS
Environmentally Preferable