CRITERION™ A-1 Medium, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7570 C7571 C7572 C7573
CA89406-404EA 234 CAD
CA89406-404 CA89406-406 CA89406-408 CA89406-410
CRITERION™ A-1 Medium, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ A-1 Medium is recommended for the detection of fecal coliforms in foods, treated wastewater, and seawater.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Certifications: BSE Free
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature –2-30°C