Mueller Hinton Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7520 C7521 C7522 C7523
CA89406-372EA 184.48 CAD
CA89406-372 CA89406-374 CA89406-376 CA89406-378
Mueller Hinton Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Mueller Hinton Broth is recommended for use in the cultivation of a wide variety of microorganisms
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Mueller Hinton Broth is also recommended for antimicrobial susceptibility testing of aerobic microorganisms by broth dilution methods. Dehydratedculture media with engineered container.
Certifications: BSE Free