m Endo LES Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7410 C7411 C7412 C7413
CA89406-332EA 247.33 CAD
CA89406-332 CA89406-334 CA89406-336 CA89406-338
m Endo LES Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ m Endo LES Agar is recommended for the enumeration of coliforms in water by membrane filtration
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free