Violet Red Bile Agar with MUG, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7250 C7251 C7252 C7253
CA89406-226EA 205.78 CAD
CA89406-226 CA89406-228 CA89406-230 CA89406-232
Violet Red Bile Agar with MUG, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Violet Red Bile Agar with MUG is primarily for use in the detection of Escherichia coli and total coliforms in food and dairy products.
CRITERION™ Dehydrated Culture Media and container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Violet Red Bile Agar with MUG is primarily for use in the detection of Escherichia coli and total coliforms in food and dairy products.
Certifications: BSE Free