Mycobiotic Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6430 C6431 C6432 C6433
CA89405-768EA 281.13 CAD
CA89405-768 CA89405-770 CA89405-772 CA89405-774
Mycobiotic Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Mycobiotic Agar is recommended for use in the selective isolation and recovery of pathogenic fungi and dermatophytes from clinical specimens; especially hair, skin, and nails
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free