Middlebrook 7H10 Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6280 C6281 C6282 C6283
CA89405-692EA 191.65 CAD
CA89405-692 CA89405-694 CA89405-696 CA89405-698
Middlebrook 7H10 Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Middlebrook 7H10 Agar Base is recommended for use in qualitative procedures for the isolation and cultivation of Mycobacteria species
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
OADC Supplement, product 89407-732, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free