CRITERION™ Malt Extract, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6210 C6211 C6212 C6213
CA89405-660EA 200.01 CAD
CA89405-660 CA89405-662 CA89405-664 CA89405-666
CRITERION™ Malt Extract, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Malt Extract is a dehydrated extract of malt for use in preparing microbiological media
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: BSE Free