Dextrose Tryptone Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5631 C5632 C5633 C5634
CA89405-396EA 300.42 CAD
CA89405-396 CA89405-398 CA89405-400 CA89405-402
Dextrose Tryptone Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Dextrose Tryptone Broth for the cultivation of thermophilic “flat sour” microorganisms related to the spoilage of food
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydrated culture media with engineered container.
Certifications: BSE Free