Azide Dextrose Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5050 C5051 C5052 C5053
CA89405-088EA 185.28 CAD
CA89405-088 CA89405-090 CA89405-092 CA89405-094
Azide Dextrose Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Azide Dextrose Broth is used to detect the presence of streptococci in water, shellfish, sewage, and milk
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free